missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TJP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29765
This item is not returnable.
View return policy
Description
TJP3 Polyclonal antibody specifically detects TJP3 in Human samples. It is validated for Immunoprecipitation
Specifications
| TJP3 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27134 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| MGC119546, Tight junction protein 3, tight junction protein 3 (zona occludens 3), ZO-3, ZO3tight junction protein ZO-3, zona occludens 3, Zona occludens protein 3, Zonula occludens protein 3 | |
| Between 325 and 375 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction