missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Proprotein Convertase 9/PCSK9 Antibody (RG7652) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28201-1mg
This item is not returnable.
View return policy
Description
Proprotein Convertase 9/PCSK9 Monoclonal antibody specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Proprotein Convertase 9/PCSK9 | |
| Monoclonal | |
| Unconjugated | |
| Q8NBP7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| RG7652 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| PCSK9 | |
| 1 mg | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 255738 | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?