missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLITRK5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30284
This item is not returnable.
View return policy
Description
SLITRK5 Polyclonal antibody specifically detects SLITRK5 in Human samples. It is validated for Western Blot
Specifications
| SLITRK5 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| bA364G4.2, FLJ58374, KIAA0918, leucine-rich repeat-containing protein 11, LRRC11, SLIT and NTRK-like family, member 5, SLIT and NTRK-like protein 5 | |
| Between 875 and 925 | |
| 100 μg | |
| Neuroscience | |
| 26050 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction