missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol, U.S.P. - F.C.C., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Unit Size:
19 L
4 L
500mL
Specifications
Specifications
| Quantity | 4 L |
| Formula Weight | 76.1 |
| Flash Point | 99.00°C |
| Grade | FCC/USP |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 1.04kg |
| Chemical Name or Material | Propylene Glycol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction