missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylthiouracil, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 6-Propyl-2-thiouracil |
| CAS | 51-52-5 |
| Analyte or Component Names | 6-Propyl-2-thiouracil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H10 N2 O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Propylthiouracil, 2,3-dihydro-6-propyl-2-thioxopyrimidin-4(1H)-one |
| InChI Formula | InChI=1S/C7H10N2OS/c1-2-3-5-4-6(10)9-7(11)8-5/h4H,2-3H2,1H3,(H2,8,9,10,11) |
| Solution Type | Neat |
| SMILES | CCCC1=CC(=O)NC(=S)N1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction