Learn More
Propyne, ca. 4% in N,N-Dimethylformamide, ca. 1.0mol/L, TRC
Product Code. 30416461
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416461 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30416461 Supplier Toronto Research Chemicals Supplier No. TRCP8359851ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Propyne
CAS 74-99-7
Molecular Formula C3H4
Quantity 1 mL
Synonym Allylene, Methylacetylene, Propine
InChI Formula InChI=1S/C3H4/c1-3-2/h1H,2H3
SMILES CC#C
IUPAC Name prop-1-yne
Molecular Weight (g/mol) 40.06
Formula Weight 40.0313
Concentration or Composition (by Analyte or Components) 4 %