missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prosapip1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556614
This item is not returnable.
View return policy
Description
Immunogen sequence: QEFAMKSVGT RTGGGGSQGS FPGPRGSGSG ASRERPGRYP SEDKGLANSL YLNGELRGSD HTDVCGNVVG SSGGSSSSGG SDKAPPQYRE PS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Prosapip1 is a binding partner of the postsynaptic density (PSD) protein ProSAP2/Shank3, a major scaffolding protein in excitatory brain synapses. Prosapip1 is thought to bind ProSAP2/Shank3 through a PDZ domain in ProSAP2/Shank3 and link it in turn to the Spine-associated Rap-Gap (SPAR). Prosapip1 shares significant homology with another PSD protein, PSD-Zip70, with both coding for a Fez1 domain that can be found in a total of four related protein, forming a novel family of scaffolding molecules termed 'Fezzins'. Recent studies show that Prosapip1 can be found in endocrine cells in which it is localized primarily in the hormone-storing compartment of these cells, suggesting that Prosapip1 may have other functions in non-neuronal cells.
Specifications
| Prosapip1 | |
| Polyclonal | |
| Unconjugated | |
| Lzts3 | |
| KIAA0552; Leucine zipper putative tumor suppressor 3; leucine zipper tumor suppressor family member 3; leucine zipper, putative tumor suppressor family member 3; LZTS3; proline rich synapse associated protein interacting protein 1; proline rich synapse associated protein interacting protein 1 (Prosapip1); ProSAP/Shank-interacting protein 1; proSAP-interacting protein 1; PROSAPIP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9762 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60299 | |
| Lzts3 | |
| Recombinant protein corresponding to Human LZTS3. Recombinant protein control fragment (Product #RP-95501). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction