Learn More
Contec™ PROSAT™ EcoCloth™
Shop All Contec Products
Click to view available options
Dimensions (L x W):
9 in. x 9 in. (23 x 23 cm)
Unit Size:
Pack of 1200
Description
- The PROSAT™ Amplitude EcoCloth™ Wipes are made using a durable, sorbent fabric that is very low in extractable residue.
- Saturated with a blend of IPA and water, these wipes are ideal for cleaning applications
Specifications
Specifications
| Length (English) | 9 in. |
| Length (Metric) | 23 cm |
| Width (English) | 9 in. |
| Width (Metric) | 23 cm |
| Material | Non-woven Polyester Cellulose |
| Disposable | Single-use |
| Dimensions (L x W) | 9 in. x 9 in. (23 x 23 cm) |
| Saturation Solution | 70% IPA, 30% DI Water |
| Sterility | Non-sterile |