missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PROSC Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99607C
This item is not returnable.
View return policy
Description
PROSC Polyclonal antibody specifically detects PROSC in Human samples. It is validated for Western Blot
Specifications
| PROSC | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| FLJ11861, proline synthase co-transcribed bacterial homolog protein, proline synthetase co-transcribed (bacterial homolog), proline synthetase co-transcribed bacterial homolog protein, proline synthetase co-transcribed homolog (bacterial) | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PROSC. | |
| 0.1 mL | |
| Signal Transduction | |
| 11212 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction