Learn More
PROSC Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99607F
This item is not returnable.
View return policy
Description
PROSC Polyclonal antibody specifically detects PROSC in Human samples. It is validated for Western Blot
Specifications
| Polyclonal | |
| Western Blot | |
| PBS | |
| Purified |