missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prostasin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA546933
This item is not returnable.
View return policy
Description
In sandwich ELISAs, less than 0.2% cross-reactivity is observed with recombinant human Prostasin. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.
Specifications
| Prostasin | |
| Polyclonal | |
| Unconjugated | |
| PRSS8 | |
| 2410039E18Rik; AI313909; C79772; CAP1; channel activating protease 1; channel-activating protease 1; fr; mCAP1; PROSTASIN; Prostasin heavy chain; Prostasin light chain; protease, serine 8; protease, serine 8 (prostasin); protease, serine, 8; protease, serine, 8 (prostasin); Prss8; Serine protease 8 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 76560 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9ESD1 | |
| PRSS8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Prostasin/Prss8 Ala30-Gln289. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction