missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostasin/Prss8 Antibody (107), Janelia Fluor™ 549 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89382JF549
This item is not returnable.
View return policy
Description
Prostasin/Prss8 Monoclonal antibody specifically detects Prostasin/Prss8 in Human samples. It is validated for ELISA
Specifications
| Prostasin/Prss8 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, PROSTASIN, protease, serine, 8, Serine protease 8 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Prostasin/Prss8 (NP_002764.1; Ala 30-Arg 322). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 107 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5652 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction