missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostate Specific Acid Phosphatase Recombinant Rabbit Monoclonal Antibody (004)

Prostate Specific Acid Phosphatase Recombinant Rabbit Monoclonal Antibody (4)
Brand: Invitrogen MA530032
This item is not returnable.
View return policy
Description
Specifications
| Prostate Specific Acid Phosphatase | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P15309 | |
| ACP3 | |
| Recombinant Human Prostatic Acid Phosphatase/ACPP protein (Met1-Lys382). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 004 | |
| Unconjugated | |
| ACP3 | |
| 5'-NT; 5'-nucleotidase; A030005E02Rik; acid phosphatase, prostate; ACP3; ACP-3; Acpp; Acpp11; EC 3.1.3.2; Ecto-5'-nucleotidase; fluoride-resistant acid phosphatase; FRAP; Lap; lysosomal acid phosphatase; PAP; PAPf39; Ppal; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; RNACPP11; Thiamine monophosphatase; TMPase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction