missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostatic Acid Phosphatase/ACPP Antibody (ACPP/1338), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54308C
This item is not returnable.
View return policy
Description
Prostatic Acid Phosphatase/ACPP Monoclonal antibody specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Prostatic Acid Phosphatase/ACPP | |
| Monoclonal | |
| DyLight 650 | |
| acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase | |
| Recombinant full-length human Prostatic Acid Phosphatase/ACPP protein (Uniprot: P15309) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 55 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| ACPP/1338 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction