Learn More
ProSwift™ C4 RP-5H Capillary LC Columns
Improve speed and resolution of LC-MS protein identification and biomarker discovery using these C4 monolithic HPLC columns.
Merk:  ThermoScientific BioLC Columns 164931
Overige gegevens : Gewicht : 0.50000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15356168
  • € 944.38 / 1 stuk
Geschatte verzenddatum
bij leverancier 12-03-2018
Toevoegen aan winkelmandje
Specificatie
Specificatie
Butyl (C4)
0.5mm
LC/MS
Poly(ethylenedimethacrylate-co-butylmethacrylate)
Monolith
1.5μm
90°C
NA
Reversed Phase
12 to 100µL/min
100mm
2000 psi (13.8 MPa)
Monolith
Most Common Organic Solvents
Analytical Column
Butyl (C4)
Video's
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.