missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PROTAC™(H-PGDS)-7

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
PROTAC™(H-PGDS)-7 is a potent Degrader of hematopoietic prostaglandin D2 synthase (H-PGDS; DC50 = 17pM after 24 hours). Comprises the H-PGDS inhibitor TFC-007 (Cat. No. 5108) directly linked to E3 ligase cereblon. Suppresses prostaglandin D2 production in vitro and in vivo. PROTAC™(H-PGDS)-8 negative control (Cat. No. 8005) also available.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-[4-[4-[[4-[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]-1-piperazinyl]carbonyl]-1-piperidinyl]phenyl]-2-phenoxy-5-pyrimidinecarboxamide |
| CAS | 2761281-50-7 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C40H38N8O7 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction