missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S 10B Antibody (OTI1G6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73627H
This item is not returnable.
View return policy
Description
Proteasome 19S 10B Monoclonal specifically detects Proteasome 19S 10B in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Proteasome 19S 10B | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PSMD2 | |
| Full length recombinant protein of human Proteasome 19S 10B (NP_002799) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1G6 | |
| Western Blot, Immunohistochemistry | |
| 26S proteasome regulatory subunit RPN1, 26S proteasome regulatory subunit S2, MGC14274,55.11 protein, P97, proteasome (prosome, macropain) 26S subunit, non-ATPase, 2, Protein 55.11, Rpn1, S226S proteasome non-ATPase regulatory subunit 2, TNFR-associated protein 2, TRAP226S proteasome subunit p97, tumor necrosis factor receptor-associated protein 2, Tumor necrosis factor type 1 receptor-associated protein 2 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway | |
| 5708.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction