missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 20S alpha 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35411-20ul
This item is not returnable.
View return policy
Description
Proteasome 20S alpha 5 Polyclonal antibody specifically detects Proteasome 20S alpha 5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| Proteasome 20S alpha 5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-241 of human Proteasome 20S alpha 5 (NP_002781.2).,, Sequence:, MFLTRSEYDRGVNTFSPEGRLFQVEYAIEAIKLGSTAIGIQTSEGVCLAVEKRITSPLMEPSSIEKIVEIDAHIGCAMSGLIADAKTLIDKARVETQNHWFTYNETMTVESVTQAVSNLALQFGEEDADPGAMSRPFGVALLFGGVDEKGPQLFHMDPSGTFVQCDARAIGSASEGAQSSLQEVYHKSMTLKEAIKSSLIILKQVMEEKLNATNIELATVQPGQNFHMFTKEELEEVIKDI | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 5686 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction