missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 20S alpha 6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87796
This item is not returnable.
View return policy
Description
Proteasome 20S alpha 6 Polyclonal antibody specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Proteasome 20S alpha 6 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EAANWKYKYGYEIPVDMLCKRIADISQVYTQNAEMRPLGCCMILIGIDEEQGPQVYKCDPAGYYCGFKATAAGVKQTESTSFLEKKVKKKFDWTFEQTVETAITCLSTVLSIDFKPSEIEVGVVTVENPKFRILTEAEIDAHL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5687 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction