missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 20S alpha 6 Antibody (OTI4C9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73637F
This item is not returnable.
View return policy
Description
Proteasome 20S alpha 6 Monoclonal specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| Proteasome 20S alpha 6 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PSMA6 | |
| Full length recombinant protein of human Proteasome 20S alpha 6 (NP_002782) produced in E.coli. | |
| 0.1 mL | |
| 5687.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry | |
| OTI4C9 | |
| Flow Cytometry, Immunohistochemistry | |
| EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction