missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 26S S5 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3319B
This item is not returnable.
View return policy
Description
Proteasome 26S S5 Polyclonal antibody specifically detects Proteasome 26S S5 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Proteasome 26S S5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Biotin | |
| KIAA007226S proteasome non-ATPase regulatory subunit 5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 5,26S proteasome subunit S5B, S5BMGC23145,26S protease subunit S5 basic | |
| Synthetic Peptide: L(485) S E G P Y Y V K P V S T T A V E G A E(504) | |
| 0.1 mL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5711 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction