missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome beta 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54583
This item is not returnable.
View return policy
Description
Proteasome beta 1 Polyclonal specifically detects Proteasome beta 1 in Human samples. It is validated for Western Blot.
Specifications
| Proteasome beta 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.4.25.1, FLJ25321, HC5, KIAA1838, Macropain subunit C5, Multicatalytic endopeptidase complex subunit C5, PMSB1, proteasome (prosome, macropain) subunit, beta type, 1, proteasome beta 1 subunit, Proteasome component C5, Proteasome gamma chain, proteasome subunit beta type-1, proteasome subunit HC5, PSC5 | |
| Rabbit | |
| 26 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: beta type-1. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P20618 | |
| PSMB1 | |
| Synthetic peptides corresponding to PSMB1(proteasome (prosome, macropain) subunit, beta type, 1) The peptide sequence was selected from the middle region of PSMB1 (NP_002784). Peptide sequence NNKAMTTGAIAAMLSTILYSRRFFPYYVYNIIGGLDEEGKGAVYSFDPVG. | |
| Affinity purified | |
| RUO | |
| 5689 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction