Learn More
Trajan Scientific and Medical™ ProteCol™ PEEK Ferrules
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Ferrules offer complete versatility for connecting capillary tubing.
Brand:  Trajan Scientific and Medical™ 213006
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15229213
  • € 40.00 / Pack of 5
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
1.59 mm to 1.59 mm
1/16 in. to 1/16 in.
ProteCol PEEK Ferrules 1/16 in. to 1/16 in.
5/Pk.