Learn More
Trajan Scientific and Medical™ ProteCol™ PEEK Ferrules
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Ferrules offer complete versatility for connecting capillary tubing.
Brand:  Trajan Scientific and Medical™ 223006
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15239223
  • € 40.00 / Pack of 5
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
0.79 mm to 0.36 mm
Stainless Steel
ProteCol PEEK Ferrules 1/32 in. to 0.36 mm