missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein C Inhibitor Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101753
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Serpin A5.
This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Protein C Inhibitor | |
| Polyclonal | |
| Unconjugated | |
| SERPINA5 | |
| 4933415L04; acrosomal serine protease inhibitor; alpha-1 antiproteinase; antitr; antitrypsin; LOC480424; LOC483954; PAI3; PAI-3; Pci; PCI-B; PLANH3; Plasma serine protease inhibitor; plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; PROCI; Protein C inhibitor; serine (or cysteine) peptidase inhibitor, clade A, member 5; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serine (or cysteine) proteinase inhibitor, clade A, member 5; serpin A5; serpin B3; serpin family A member 5; serpin family B member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serpin peptidase inhibitor, clade A, member 5; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINA5; SERPINB3; SERPINB4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 268591, 5104, 65051 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05154, P70458 | |
| SERPINA5 | |
| A synthesized peptide derived from human SERPINA5(Accession P05154), corresponding to amino acid residues G310-T360. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction