missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Kinase A regulatory subunit I alpha Antibody (OTI6C7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73659UV
This item is not returnable.
View return policy
Description
Protein Kinase A regulatory subunit I alpha Monoclonal specifically detects Protein Kinase A regulatory subunit I alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Protein Kinase A regulatory subunit I alpha | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PRKAR1A | |
| Full-length protein expressed in 293T cell transfected with human Protein Kinase A regulatory subunit I alpha expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6C7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| cAMP-dependent protein kinase regulatory subunit RIalpha, cAMP-dependent protein kinase type I-alpha regulatory chain, cAMP-dependent protein kinase type I-alpha regulatory subunit, CAR, CNC, CNC1, MGC17251, PKA R1 alpha, PKA1, PKR1, PPNAD1, PRKAR1, protein kinase A type 1a regulatory subunit, protein kinase, cAMP-dependent, regulatory, type I, alpha (tissue specificextinguisher 1), Tissue-specific extinguisher 1, TSE1DKFZp779L0468 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 5573.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction