Learn More
Thermo Scientific™ Protein Kinase C-delta Peptide Substrate
Product Code. 15497289
Click to view available options
Substrate Quantity:
0.5mg
1mg
Unit Size:
0.5mg
1mg
Product Code. 15497289
Brand: Thermo Scientific™ J66073.M^
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For cellular and molecular biology applications
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Color White to off-white
Form Lyophilized
Molecular Weight (g/mol) 2487.99
Storage Requirements -30° to -10°C
Substrate Quantity 1mg