missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein kinase-like protein SgK493 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80756
This item is not returnable.
View return policy
Description
Protein kinase-like protein SgK493 Polyclonal specifically detects Protein kinase-like protein SgK493 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Protein kinase-like protein SgK493 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q504Y2 | |
| PKDCC | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:STDCILEFPARNFTLPCSAQGWCEGMNEKRNLYNAYRFFFTYLLPHSAPPSLRPLLDSIVNATGELAWGVDETLAQLEKVLHLYRSGQ | |
| 0.1 mL | |
| Protein Kinase | |
| 91461 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 2.7, FLJ18197, MGC125960, protein kinase domain containing, cytoplasmic homolog (mouse), protein kinase domain-containing protein, cytoplasmic, Protein kinase-like protein SgK493, SgK493, Sugen kinase 493, Vertebrate lonesome kinase, Vlk | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction