missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein O-Fucosyltransferase 1/POFUT1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76940MFV450
This item is not returnable.
View return policy
Description
Protein O-Fucosyltransferase 1/POFUT1 Polyclonal antibody specifically detects Protein O-Fucosyltransferase 1/POFUT1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Protein O-Fucosyltransferase 1/POFUT1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FUT12EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 1, KIAA0180MGC2482, o-fucosyltransferase protein, O-Fuc-T, O-FucT-1, O-FUT, Peptide-O-fucosyltransferase 1, protein O-fucosyltransferase 1 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human POFUT1. The immunogen is located within amino acids 90 - 140 of POFUT1. Amino Acid Squence: DFMEKLAPTHWPPEKR | |
| 0.1 mL | |
| Signal Transduction | |
| 23509 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction