missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Protein Phosphatase inhibitor 1 Antibody (SR3103), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 339.00 - € 406.00
Specifications
| Antigen | Protein Phosphatase inhibitor 1 |
|---|---|
| Clone | SR3103 |
| Dilution | Western Blot 1:5000-1:20000, Flow Cytometry 1:20, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041806
|
Novus Biologicals™
NBP3-27703-100ul |
100 μL |
€ 406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041657
|
Novus Biologicals™
NBP3-27703-50ul |
50 μL |
€ 339.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein Phosphatase inhibitor 1 Monoclonal antibody specifically detects Protein Phosphatase inhibitor 1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Protein Phosphatase inhibitor 1 | |
| Western Blot 1:5000-1:20000, Flow Cytometry 1:20, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| I1, I-1, IPP-1, IPP1inhibitor-1, protein phosphatase 1 regulatory subunit 1A, protein phosphatase 1, regulatory (inhibitor) subunit 1A, Protein phosphatase inhibitor 1, protein phosphatase inhibitor-1 | |
| A synthesized peptide derived from human Protein Phosphatase inhibitor 1 (Uniprot #: Q13522) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3103 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 5502 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title