missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Z Antibody (OTI4D5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73673B
This item is not returnable.
View return policy
Description
Protein Z Monoclonal specifically detects Protein Z in Human samples. It is validated for Immunohistochemistry.
Specifications
| Protein Z | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PROZ | |
| Recombinant protein fragment corresponding to amino acids 115-400 of human Protein Z (NP_003882) produced in E.coli. | |
| 0.1 mL | |
| 8858.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI4D5 | |
| Immunohistochemistry | |
| protein Z, vitamin K-dependent plasma glycoprotein, PZ, vitamin K-dependent protein Z | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction