missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteinase 3/Myeloblastin/PRTN3 Antibody (SR1444), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22274-100ul
This item is not returnable.
View return policy
Description
Proteinase 3/Myeloblastin/PRTN3 Monoclonal antibody specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Proteinase 3/Myeloblastin/PRTN3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1444 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| ACPA, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen | |
| A synthesized peptide derived from human Proteinase 3/Myeloblastin/PRTN3 (Uniprot #: P24158) | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 5657 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction