missing translation for 'onlineSavingsMsg'
Learn More

Prothioconazole, TRC

Product Code. 30416594
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416594 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416594 Supplier Toronto Research Chemicals Supplier No. TRCP838830250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prothioconazole
CAS 178928-70-6
Purity Grade Notes HPLC
Molecular Formula C14 H15 Cl2 N3 O S
Quantity 250 mg
Synonym 2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-2,4-dihydro-3H-1,2,4-triazole-3-thione, JAU 6476, Joao, Proline (fungicide), Proline 275, Proline 480SC, Prothioconazole, Redigo, Rudis
InChI Formula InChI=1S/C14H15Cl2N3OS/c15-11-4-2-1-3-10(11)7-14(20,13(16)5-6-13)8-19-12(21)17-9-18-19/h1-4,9,20H,5-8H2,(H,17,18,21)
SMILES OC(CN1NC=NC1=S)(Cc2ccccc2Cl)C3(Cl)CC3
IUPAC Name 2-[2-(1-chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1H-1,2,4-triazole-3-thione
Molecular Weight (g/mol) 344.26
Formula Weight 343.0313
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.