missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prothrombin Monoclonal Antibody (C10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541568
This item is not returnable.
View return policy
Description
Prothrombin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
Specifications
| Prothrombin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P18292 | |
| F2 | |
| Recombinant protein Coagulation Factor II. The antigen corresponds to amino acid range Ser201- Arg323 of the target protein. | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C10 | |
| Unconjugated | |
| F2 | |
| Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; Cf2; Cf-2; coagulation factor 2; coagulation factor II; coagulation factor II (thrombin); coagulation factor II [thrombin]; coagulation factor II, thrombin; F2; FII; kringle-2 domain; fragment-2; prepro-coagulation factor II; preprothrombin; prothrombin; prothrombin B-chain; prothrombin precursor; PT; RPRGL2; serine protease; THPH1; THROMBIN; Thrombin heavy chain; Thrombin light chain | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 29251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction