missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Prothrombin Monoclonal Antibody (C13)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA541567

 View more versions of this product

Product Code. 17858082

  • € 489.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Prothrombin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prothrombin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P18292
F2
Recombinant protein Coagulation Factor II. The antigen corresponds to amino acid range 201-323 of the target protein.
200 μL
Primary
Rat
Antibody
IgG2b κ
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
C13
Unconjugated
F2
Activation peptide fragment 1; Activation peptide fragment 2; bovine prothrombin protein; Cf2; Cf-2; coagulation factor 2; coagulation factor II; coagulation factor II (thrombin); coagulation factor II [thrombin]; coagulation factor II, thrombin; F2; FII; kringle-2 domain; fragment-2; prepro-coagulation factor II; preprothrombin; prothrombin; prothrombin B-chain; prothrombin precursor; PT; RPRGL2; serine protease; THPH1; THROMBIN; Thrombin heavy chain; Thrombin light chain
Mouse
Protein A/G
RUO
29251
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.