Learn More
Macherey-Nagel Bioanalysis™ Protino™ Ni-NTA 5 mL FPLC™ Columns
The method of choice for His-tag protein purification in a ready-to-use 5 mL FPCL columns.
Brand:  Macherey-Nagel Bioanalysis™ 745415.5
Additional Details : Weight : 0.18400kg
 View more versions of this product
Product Code. 11982522
  • € 460.00 / Pack of 5
Estimated Shipment: 17-08-2021
Add to basket
Description
Description
  • Target: Protein
  • Technology: Immobilized metal ion affinity chromatography
  • Matrix: 6% beaded agarose (cross-linked), precharged with Ni2+
  • Bead size: 45 to 165 μm
  • Binding capacity: 250 mg
  • Enable fast and convenient purification of recombinant polyhistidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC)
  • Proteins from any expression system can be purified under native or denaturing
Specifications
Specifications
Protein solution
NTA (nitrilotriacetic acid)
Store at 4 to 8°C
Purification of His-tag proteins
5 x 5 mL
Videos
Safety and Handling
Safety and Handling
Hazard Category
  • Respiratory sensitiser Category 1