Learn More
Protocadherin-1 Antibody (648127), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5899Y
This item is not returnable.
View return policy
Description
Protocadherin-1 Monoclonal antibody specifically detects Protocadherin-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Protocadherin-1 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5097 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 648127 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| cadherin-like 1, Cadherin-like protein 1, FLJ53887, MGC45991, pc42, PC42PCDH42, protocadherin 1, protocadherin 1 (cadherin-like 1), protocadherin 42, protocadherin-1, protocadherin-42 | |
| E. coli-derived recombinant human Protocadherin-1, Thr58-Asn162, Accession # Q08174 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |