missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocadherin gamma C3 Antibody (926518) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB83641JF635
This item is not returnable.
View return policy
Description
Protocadherin gamma C3 Monoclonal antibody specifically detects Protocadherin gamma C3 in Human samples. It is validated for Flow Cytometry
Specifications
| Protocadherin gamma C3 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin gamma C3, Met1-Tyr693, Accession # Q9UN70 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 926518 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 5098 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction