missing translation for 'onlineSavingsMsg'
Learn More

Protocatechuic acid, MedChemExpress

Product Code. 16411506
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411506 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411506 Supplier Medchem Express Supplier No. HYN0294
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Protocatechuic acid is a phenolic compound which exhibits neuroprotective effect.

Protocatechuic acid is a phenolic compound which exhibits neuroprotective effect.

Specifications

Chemical Name or Material Protocatechuic acid
CAS 99-50-3
Color White
Purity Grade Notes Research
Molecular Formula C7H6O4
Quantity 200 mg
Synonym 3,4-Dihydroxybenzoic acid
Solubility Information DMSO : 100 mg/mL (648.85 mM; Need ultrasonic) ∣H2O : 10 mg/mL (64.88 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 154.12
Formula Weight 154.12
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.