missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protogenin Antibody (OTI2B3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73674IR
This item is not returnable.
View return policy
Description
Protogenin Monoclonal specifically detects Protogenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Protogenin | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PRTG | |
| Full length recombinant protein of human Protogenin (NP_776175) produced in HEK293T cells. | |
| 0.1 mL | |
| 283659.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2B3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), shen-dan | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction