Learn More
Protopine N-Oxide, Mikromol™
Description
Protopine N-Oxide
Specifications
Specifications
| Chemical Name or Material | Protopine N-Oxide |
| CAS | 87264-51-5 |
| Analyte or Component Names | Protopine N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H19 N O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Bis[1,3]benzodioxolo[4,5-c:5',6'-g]azecin-13(5H)-one, 4,6,7,14-tetrahydro-5-methyl-, 5-oxide, Protopine N-oxide |
| InChI Formula | InChI=1S/C20H19NO6/c1-21(23)5-4-13-7-18-19(26-10-25-18)8-14(13)16(22)6-12-2-3-17-20(15(12)9-21)27-11-24-17/h2-3,7-8H,4-6,9-11H2,1H3 |
| SMILES | C[N+]1([O-])CCc2cc3OCOc3cc2C(=O)Cc4ccc5OCOc5c4C1 |
| Molecular Weight (g/mol) | 369.37 |
| Show More |