Learn More
Prox1 Antibody (5G10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-30045AF647
Product Code. 18750454
  • ONLINE EXCLUSIVE PRICE
    € 642.00
    € 606.90 / 0.10mL
    Save € 35.10
    5% Off
Description
Prox1 Monoclonal specifically detects Prox1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prox1
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat, Chicken
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
5G10
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount
Q92786
PROX1
The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786]
0.1 mL
Vision
5629
Store at 4C in the dark.