Learn More
Prox1 Antibody (CL13472) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24560AF350
Product Code. 30302751
  • € 737.00 / 0.10mL
Description
Prox1 Monoclonal antibody specifically detects Prox1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prox1
Monoclonal
Alexa Fluor 350
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1
This antibody was developed using a recombinant protein derived from Q92786, with the exact immunogen sequence remaining proprietary.
0.1 mL
Vision
5629
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13472