Learn More
Prox1 Antibody (CL13474) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24561MFV450
Product Code. 30295089
  • € 759.00 / 0.10mL
Description
Prox1 Monoclonal antibody specifically detects Prox1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prox1
Monoclonal
mFluor Violet 450 SE
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1
This antibody was developed using a recombinant protein derived from Q92786, with the exact immunogen sequence remaining proprietary.
0.1 mL
Vision
5629
Store at 4⪚C in the dark.
IgG1
Immunocytochemistry/Immunofluorescence
CL13474
50mM Sodium Borate
Mouse
Protein A purified