missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PROX1 Recombinant Rabbit Monoclonal Antibody (9I9A7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535749
This item is not returnable.
View return policy
Description
Immunogen sequence: MPDHDSTALL SRQTKRRRVD IGVKRTVGTA SAFFAKARAT FFSAMNPQGS EQDVEYSVVQ HADGEKSNVL RKLLKRANSY EDAMMPFPGA TIISQLLKNN.
The expression pattern of the Prox1 homeo box gene suggests that it has a role in a variety of embryonic tissues, including lens. Analysis of mRNA reveals that Prox mRNA is present in many different human tissues and that lens demonstrated the highest level. Homozygous Prox1-null mice die at midgestation from multiple developmental defects, and a targeted effect on lens development has been reported. Prox1 inactivation caused abnormal cellular proliferation, downregulated expression of the cell cycle inhibitors Cdkn1b and Cdkn1c, misexpression of E-cadherin, and excessive apoptosis. Consequently, mutant lens cells failed to polarize and elongate properly, resulting in a hollow lens. The Prox1 gene is expressed in a subpopulation of endothelial cells that by budding and sprouting give rise to the lymphatic system. Prox1 appears to be a specific and required regulator of the development of the lymphatic system. Prox1 also has been document to be required for hepatocyte migration in the mouse. Loss of Prox1 results in a smaller liver with a reduced population of clustered hepatocytes. The homeodomain protein Prox1 regulates the egress of progenitor cells from the cell cycle in the embryonic mouse retina. Cells lacking Prox1 are less likely to stop dividing, and ectopic expression of Prox1 forces progenitor cells to exit the cell cycle. Prox1 acts as a key participant in progenitor-cell proliferation and cell-fate determination in the vertebrate retina.
Specifications
| PROX1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48437, Q92786 | |
| Prox1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PROX1 (Q92786). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 9I9A7 | |
| Unconjugated | |
| Prox1 | |
| A230003G05Rik; homeobox prospero-like protein PROX1; prospero homeobox 1; prospero homeobox protein 1; prospero-related homeobox 1; Prox 1; PROX1; PROX-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19130, 305066, 5629 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction