Learn More
PROX1 Mouse anti-Human, Unconjugated, Clone: 1D4B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67438-1-IG-20UL
 View more versions of this product
Product Code. 16880066
  • € 145.00 / 20µL
Description
The PROX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PROX1. This antibody recognizes human, rat, mouse antigen. The PROX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PROX1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1D4B1
Unconjugated
20 μL
5629
IgG2b