missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PROX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA568903
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This target displays homology in the following species: Mouse: 100%; Rat: 79%.
Transcription regulator. Does not seem to be essential for embryonic development and postnatal survival.
Specifications
| PROX2 | |
| Polyclonal | |
| Unconjugated | |
| PROX2 | |
| 1700058C01Rik; Homeobox prospero-like protein PROX2; prospero homeobox 2; prospero homeobox protein 2; PROX2; PROX-2; RGD1310315 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 73422 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BII1 | |
| PROX2 | |
| synthetic peptide directed towards the n terminal region of mouse PROX2. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction