Learn More
PROZ Monoclonal Antibody (OTI4D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806759S
This item is not returnable.
View return policy
Description
PROZ Monoclonal antibody specifically detects PROZ in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PROZ | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20138 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 945 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| OTI4D5 | |
| Unconjugated | |
| CD33 | |
| protein Z, vitamin K dependent plasma glycoprotein; protein Z, vitamin K-dependent plasma glycoprotein; PROZ; PZ; vitamin K-dependent protein Z; vitamin K-dependent protein Z precursor variant 1 | |
| Full length human recombinant protein of human CD33 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |