Learn More
Invitrogen™ PrP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596175
Description
Immunogen sequence: KKRPKPGGWN TGGSRYPGQG SPGGNRYPPQ GGGGWGQPHG GGWGQPHGGG WGQPHGGGWG QPHGGGWGQG GGTHSQWNKP SKPKTNMKHM AGAAAAGAVV GGLGGYMLGS AMSRPIIHFG SDYEDRYYRE NMHRYPNQVY YRPMDEYSNQ NNFVHDCVNI TIKQHTVTTT TKGENFTETD VKMMERVVEQ MCITQYERES QAYYQRGS; Positive Samples: Mouse brain; Cellular Location: Cell membrane, Cytoplasm, GPI-anchor, Golgi apparatus, Lipid-anchor, Nucleus.
Human prion protein (PrP), also known as PRNP, is a ubiquitously expressed GPI-anchored cell surface glycoprotein associating with lipid raft components and functioning as a signaling molecule. PrP plays a role in apoptosis in a cell context-dependent manner, is involved in proliferation of epithelial cells and in distribution of junction-associated proteins in human enterocytes. Conversion of this normal cellular prion protein (PrPc) into an abnormal conformer (PrPSc) is the crucial step associated with triggering the pathogenesis of the prion neurodegenerative disorders, such as the Creutzfeld-Jakob disease (CJD). Whereas PrPc is rich in alpha-helices, the PrPSc form has higher content of beta-sheets and is resistant to proteinase K.
Specifications
| PrP | |
| Polyclonal | |
| Unconjugated | |
| PRNP | |
| AA960666; AI325101; Alternative prion protein; alternative prion protein; major prion protein; AltPrP; ASCR; BSE; CD230; CD230 antigen; CJD; GSS; KURU; major prion protein; major prion protein; alternative prion protein; major scrapie-associated fibril protein 1; MGC26679; p27-30; PK resistant core; prion protein; prion protein (p27-30) (Creutzfeldt-Jakob disease, Gerstmann-Strausler-Scheinker syndrome, fatal familial insomnia); prion protein precursor; prion protein precursor PrP; prion protein PrP; prion protein variant a; prion protein variant b; prion protein, PrP; prion protein, structural; prion-related protein; PRIP; Prn; Prn-i; Prnp; Prn-p; PrP; PrP>C>; PrP27-30; PrP33-35C; PrPc; PrPSc; Sinc; SIP; TSE | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19122, 5621 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F7VJQ1, P04156, P04925 | |
| PRNP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-230 of human Prion Protein (NP_0010735921). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.