missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRP6 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA530496

Product Code. 13404997

  • € 520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (89%), Xenopus laevis (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRP6
Polyclonal
Unconjugated
PRPF6
1190003A07Rik; 2610031L17Rik; androgen receptor N-terminal domain transactivating protein-1; androgen receptor N-terminal domain-transactivating protein 1; ANT1; ANT-1; C20orf14; hPrp6; p102 U5 small nuclear ribonucleoprotein particle-binding protein; pre-mRNA processing factor 6; pre-mRNA-processing factor 6; Prp6; PRP6 homolog; PRP6 pre-mRNA processing factor 6 homolog; PRP6 pre-mRNA splicing factor 6 homolog; PRP6 pre-mRNA splicing factor 6 homolog (yeast); Prpf6; putative mitochondrial outer membrane protein import receptor; RGD1307103; RP60; SNRNP102; TOM; U5 snRNP-associated 102 kDa protein; U5-102 kDa protein; U5-102K
Rabbit
Antigen affinity chromatography
RUO
24148
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.8 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O94906
PRPF6
Recombinant protein encompassing a sequence within the center region of human PRP6. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.